Assay Details
Gene Symbol: AtHB23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102460.4 | NP_564268.1 | 1 - 2 | 404 | 80 | |
GenBank mRNA | AK228056.1 | - | 1 - 2 | 212 | 80 | |
AY084913.1 | - | 1 - 2 | 213 | 80 | ||
BT009727.1 | - | 1 - 2 | 105 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
