Assay Details
Gene Symbol: NTF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084143.2 | NP_001077612.1 | 1 - 2 | 414 | 61 | |
NM_102562.4 | NP_174118.1 | 1 - 2 | 516 | 61 | ||
GenBank mRNA | BT025598.1 | - | 1 - 2 | 304 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
