Assay Details
Gene Symbol: SYP61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102617.4 | NP_564310.1 | 3 - 4 | 357 | 75 | |
GenBank mRNA | AF355754.2 | - | 2 - 3 | 161 | 75 | |
AY150420.1 | - | 2 - 3 | 161 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
