Assay Details
Gene Symbol: SYP61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036025.5 | NP_001031102.2 | 6 - 7 | 639 | 69 | |
NM_001332815.1 | NP_001321010.1 | 3 - 4 | 511 | 69 | ||
NM_102617.4 | NP_564310.1 | 6 - 7 | 757 | 69 | ||
GenBank mRNA | AF355754.2 | - | 5 - 6 | 561 | 69 | |
AY150420.1 | - | 5 - 6 | 561 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
