Assay Details
| Gene Symbol: AT1G29025 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198180.2 | NP_001185109.1 | 6 - 7 | 1410 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G29025| Gene Symbol: AT1G29025 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198180.2 | NP_001185109.1 | 6 - 7 | 1410 | 63 | |
Set Membership: |
|