Assay Details
Gene Symbol: UGE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334071.1 | NP_001321118.1 | 4 - 5 | 707 | 90 | |
NM_104996.6 | NP_564811.1 | 4 - 5 | 756 | 90 | ||
GenBank mRNA | AK117913.1 | - | 4 - 5 | 539 | 90 | |
AY085887.1 | - | 4 - 5 | 684 | 90 | ||
BT024882.1 | - | 4 - 5 | 488 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
