Assay Details
Gene Symbol: BEE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198465.1 | NP_001185394.1 | 1 - 2 | 431 | 134 | |
NM_106043.3 | NP_177524.2 | 1 - 2 | 463 | 134 | ||
GenBank mRNA | AK118781.1 | - | 1 - 2 | 429 | 134 | |
BT006055.1 | - | 1 - 2 | 377 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
