Assay Details
Gene Symbol: BEE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106043.3 | NP_177524.2 | 5 - 6 | 787 | 83 | |
GenBank mRNA | AK118781.1 | - | 5 - 6 | 753 | 83 | |
BT006055.1 | - | 5 - 6 | 701 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
