Assay Details
Gene Symbol: SS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106059.5 | NP_177540.3 | 1 - 2 | 374 | 79 | |
GenBank mRNA | AK230236.1 | - | 266 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
