Assay Details
Gene Symbol: SS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106061.4 | NP_177542.1 | 1 - 2 | 400 | 65 | |
GenBank mRNA | AY034970.1 | - | 1 - 2 | 266 | 65 | |
AY062976.1 | - | 1 - 2 | 234 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
