Assay Details
Gene Symbol: SS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106061.4 | NP_177542.1 | 2 - 3 | 671 | 61 | |
GenBank mRNA | AY034970.1 | - | 2 - 3 | 537 | 61 | |
AY062976.1 | - | 2 - 3 | 505 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
