Assay Details
| Gene Symbol: GR-RBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106083.5 | NP_177563.1 | 3 - 4 | 357 | 86 | |
| GenBank mRNA | BT002037.1 | - | 3 - 4 | 253 | 86 | |
| BT006315.1 | - | 2 - 3 | 184 | 86 | ||
More Information
Set Membership: |
|

GR-RBP5
Genomic Map