Assay Details
Gene Symbol: NF-YC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122673.2 | NP_198143.1 | 1 | 664 | 76 | |
GenBank mRNA | DQ056694.1 | - | 1 | 516 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
