Assay Details
| Gene Symbol: AT5G28050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036882.1 | NP_001031959.1 | 4 - 5 | 460 | 98 | |
| NM_001344057.1 | NP_001330272.1 | 3 - 4 | 604 | 98 | ||
| NM_122688.4 | NP_198157.1 | 3 - 4 | 536 | 98 | ||
| GenBank mRNA | AY039873.1 | - | 3 - 4 | 325 | 98 | |
| AY133663.1 | - | 3 - 4 | 260 | 98 | ||
More Information
Set Membership: |
|

AT5G28050
Genomic Map