Assay Details
Gene Symbol: GS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036892.1 | NP_001031969.1 | 5 - 6 | 657 | 105 | |
NM_001085170.1 | NP_001078639.1 | 5 - 6 | 646 | 105 | ||
NM_122954.4 | NP_198413.1 | 5 - 6 | 871 | 105 | ||
GenBank mRNA | AF428319.1 | - | 5 - 6 | 651 | 105 | |
AF428461.1 | - | 5 - 6 | 651 | 105 | ||
AK317110.1 | - | 5 - 6 | 658 | 105 | ||
AY081252.1 | - | 5 - 6 | 605 | 105 | ||
AY088222.1 | - | 5 - 6 | 684 | 105 | ||
AY091114.1 | - | 5 - 6 | 653 | 105 | ||
AY122977.1 | - | 4 - 5 | 547 | 105 | ||
S69727.1 | - | 5 - 6 | 618 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
