Assay Details
| Gene Symbol: AT5G38640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123225.3 | NP_198680.1 | 2 - 3 | 662 | 84 | |
| GenBank mRNA | AY062523.1 | - | 2 - 3 | 662 | 84 | |
| BT008788.1 | - | 2 - 3 | 612 | 84 | ||
More Information
Set Membership: |
|

AT5G38640
Genomic Map