Assay Details
Gene Symbol: ABCB27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344283.1 | NP_001332027.1 | 5 - 6 | 531 | 78 | |
NM_123266.8 | NP_198720.2 | 5 - 6 | 595 | 78 | ||
GenBank mRNA | AK229810.1 | - | 5 - 6 | 477 | 78 | |
DQ469798.1 | - | 5 - 6 | 395 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
