Assay Details
| Gene Symbol: ABCB27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344283.1 | NP_001332027.1 | 9 - 10 | 1032 | 65 | |
| NM_123266.8 | NP_198720.2 | 9 - 10 | 1096 | 65 | ||
| GenBank mRNA | AK229810.1 | - | 9 - 10 | 978 | 65 | |
| DQ469798.1 | - | 9 - 10 | 896 | 65 | ||
More Information
Set Membership: |
|

ABCB27
Genomic Map