Assay Details
| Gene Symbol: PTEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123302.4 | NP_198756.2 | 2 - 3 | 407 | 94 | |
| GenBank mRNA | AJ490172.1 | - | 2 - 3 | 301 | 94 | |
| AK118115.1 | - | 2 - 3 | 389 | 94 | ||
| BT005560.1 | - | 1 - 2 | 271 | 94 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 86 and 100  Detect Genomic DNA  Arabidopsis thaliana  Probe spans exons | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 PTEN1
PTEN1 Genomic Map
Genomic Map Back To Top
 Back To Top