Assay Details
| Gene Symbol: SGR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123313.5 | NP_198767.1 | 2 - 3 | 379 | 93 | |
| GenBank mRNA | AY070486.1 | - | 2 - 3 | 229 | 93 | |
| AY091707.1 | - | 1 - 2 | 87 | 93 | ||
More Information
Set Membership: |
|

SGR4
Genomic Map