Assay Details
Gene Symbol: NAC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123323.3 | NP_198777.1 | 2 - 3 | 734 | 122 | |
GenBank mRNA | AY056327.1 | - | 2 - 3 | 596 | 122 | |
AY091191.1 | - | 2 - 3 | 473 | 122 | ||
AY174163.1 | - | 2 - 3 | 473 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
