Assay Details
| Gene Symbol: SIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123403.4 | NP_198855.1 | 2 - 3 | 1513 | 76 | |
| GenBank mRNA | AY062781.1 | - | 2 - 3 | 1373 | 76 | |
| AY081645.1 | - | 2 - 3 | 1242 | 76 | ||
More Information
Set Membership: |
|

SIP1
Genomic Map