Assay Details
| Gene Symbol: RPL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123460.5 | NP_198911.1 | 2 - 3 | 485 | 100 | |
| GenBank mRNA | AY046039.1 | - | 2 - 3 | 459 | 100 | |
| AY117267.1 | - | 2 - 3 | 401 | 100 | ||
More Information
Set Membership: |
|

RPL27
Genomic Map