Assay Details
| Gene Symbol: UVH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036914.1 | NP_001031991.1 | 1 - 2 | 902 | 157 | |
| NM_001344387.1 | NP_001330263.1 | 1 - 2 | 1368 | 157 | ||
| NM_001344388.1 | NP_001330264.1 | 1 - 2 | 902 | 157 | ||
| NM_123480.4 | NP_198931.1 | 1 - 2 | 1368 | 157 | ||
| GenBank mRNA | AF089003.1 | - | 1 - 2 | 870 | 157 | |
| AF160500.1 | - | 1 - 2 | 964 | 157 | ||
| AY140076.1 | - | 1 - 2 | 902 | 157 | ||
More Information
Set Membership: |
|

UVH1
Genomic Map