Assay Details
| Gene Symbol: UVH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036914.1 | NP_001031991.1 | 4 - 5 | 1358 | 71 | |
| NM_001344387.1 | NP_001330263.1 | 4 - 5 | 1824 | 71 | ||
| NM_001344388.1 | NP_001330264.1 | 4 - 5 | 1358 | 71 | ||
| NM_123480.4 | NP_198931.1 | 4 - 5 | 1824 | 71 | ||
| GenBank mRNA | AF089003.1 | - | 4 - 5 | 1326 | 71 | |
| AF160500.1 | - | 4 - 5 | 1420 | 71 | ||
| AY140076.1 | - | 4 - 5 | 1358 | 71 | ||
More Information
Set Membership: |
|

UVH1
Genomic Map