Assay Details
| Gene Symbol: UVH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344387.1 | NP_001330263.1 | 5 - 6 | 2566 | 97 | |
| NM_123480.4 | NP_198931.1 | 5 - 6 | 2566 | 97 | ||
| GenBank mRNA | AF089003.1 | - | 5 - 6 | 2068 | 97 | |
| AF160500.1 | - | 5 - 6 | 2162 | 97 | ||
| AY140076.1 | - | 5 - 6 | 2100 | 97 | ||
More Information
Set Membership: |
|

UVH1
Genomic Map