Assay Details
| Gene Symbol: UBC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344399.1 | NP_001331159.1 | 1 - 2 | 283 | 71 | |
| NM_123499.5 | NP_568589.1 | 1 - 2 | 283 | 71 | ||
| GenBank mRNA | AY065360.1 | - | 1 - 2 | 224 | 71 | |
| AY096587.1 | - | 1 - 2 | 50 | 71 | ||
| DQ027019.1 | - | 1 - 2 | 50 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001344399.1 transcripts.
Set Membership: |
|

UBC4
Genomic Map