Assay Details
| Gene Symbol: UBC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344399.1 | NP_001331159.1 | 2 - 3 | 853 | 146 | |
| NM_123499.5 | NP_568589.1 | 3 - 4 | 470 | 146 | ||
| GenBank mRNA | AY065360.1 | - | 3 - 4 | 411 | 146 | |
| AY096587.1 | - | 3 - 4 | 237 | 146 | ||
| DQ027019.1 | - | 3 - 4 | 237 | 146 | ||
More Information
Set Membership: |
|

UBC4
Genomic Map