Assay Details
| Gene Symbol: GLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123512.3 | NP_198963.2 | 6 - 7 | 1046 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLA1| Gene Symbol: GLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123512.3 | NP_198963.2 | 6 - 7 | 1046 | 89 | |
Set Membership: |
|