Assay Details
Gene Symbol: ABIL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344431.1 | NP_001330354.1 | 1 - 2 | 224 | 117 | |
NM_001344432.1 | NP_001330353.1 | 1 - 2 | 193 | 117 | ||
NM_123568.5 | NP_199018.1 | 1 - 2 | 305 | 117 | ||
GenBank mRNA | AK175528.1 | - | 1 - 2 | 137 | 117 | |
AK175563.1 | - | 1 - 2 | 106 | 117 | ||
AK176699.1 | - | 1 - 2 | 193 | 117 | ||
AK176912.1 | - | 1 - 2 | 101 | 117 | ||
AY817015.1 | - | 1 - 2 | 97 | 117 | ||
BT024503.1 | - | 1 - 2 | 97 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
