Assay Details
Gene Symbol: ABIL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344431.1 | NP_001330354.1 | 4 - 5 | 558 | 114 | |
NM_001344432.1 | NP_001330353.1 | 4 - 5 | 527 | 114 | ||
NM_001344433.1 | NP_001330352.1 | 2 - 3 | 339 | 114 | ||
NM_123568.5 | NP_199018.1 | 4 - 5 | 639 | 114 | ||
GenBank mRNA | AK175528.1 | - | 4 - 5 | 471 | 114 | |
AK175563.1 | - | 4 - 5 | 440 | 114 | ||
AK176699.1 | - | 4 - 5 | 527 | 114 | ||
AK176912.1 | - | 4 - 5 | 435 | 114 | ||
AY817015.1 | - | 4 - 5 | 431 | 114 | ||
BT024503.1 | - | 4 - 5 | 431 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
