Assay Details
Gene Symbol: ABIL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344431.1 | NP_001330354.1 | 6 - 7 | 656 | 85 | |
NM_001344433.1 | NP_001330352.1 | 4 - 5 | 437 | 85 | ||
NM_123568.5 | NP_199018.1 | 6 - 7 | 737 | 85 | ||
GenBank mRNA | AK175528.1 | - | 6 - 7 | 569 | 85 | |
AK175563.1 | - | 6 - 7 | 538 | 85 | ||
AK176699.1 | - | 6 - 7 | 625 | 85 | ||
AK176912.1 | - | 6 - 7 | 533 | 85 | ||
AY817015.1 | - | 6 - 7 | 529 | 85 | ||
BT024503.1 | - | 6 - 7 | 529 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
