Assay Details
Gene Symbol: AT5G42060 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123571.3 | NP_199021.1 | 1 | 509 | 71 | |
GenBank mRNA | AY072311.1 | - | 1 | 509 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
