Assay Details
| Gene Symbol: ARC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344453.1 | NP_001331076.1 | 2 - 3 | 870 | 72 | |
| NM_123613.4 | NP_199063.1 | 2 - 3 | 931 | 72 | ||
| GenBank mRNA | AY091075.1 | - | 2 - 3 | 845 | 72 | |
| AY150490.1 | - | 2 - 3 | 732 | 72 | ||
More Information
Set Membership: |
|

ARC6
Genomic Map