Assay Details
| Gene Symbol: ARC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344453.1 | NP_001331076.1 | 4 - 5 | 2121 | 87 | |
| NM_123613.4 | NP_199063.1 | 4 - 5 | 2182 | 87 | ||
| GenBank mRNA | AY091075.1 | - | 4 - 5 | 2096 | 87 | |
| AY150490.1 | - | 4 - 5 | 1983 | 87 | ||
More Information
Set Membership: |
|

ARC6
Genomic Map