Assay Details
| Gene Symbol: PAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203528.2 | NP_001190457.1 | 4 - 5 | 2499 | 103 | |
| NM_001344465.1 | NP_001330498.1 | 4 - 5 | 2467 | 103 | ||
| NM_123652.3 | NP_199101.1 | 4 - 5 | 2514 | 103 | ||
More Information
Set Membership: |
|

PAH2
Genomic Map