Assay Details
Gene Symbol: COP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123663.3 | NP_199111.1 | 12 - 13 | 1281 | 67 | |
GenBank mRNA | AF395060.1 | - | 12 - 13 | 1114 | 67 | |
AK176360.1 | - | 12 - 13 | 1196 | 67 | ||
BT025548.1 | - | 12 - 13 | 1114 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
