Assay Details
Gene Symbol: PUM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123677.2 | NP_199125.2 | 1 - 2 | 861 | 109 | |
GenBank mRNA | DQ056705.1 | - | 1 - 2 | 861 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
