Assay Details
Gene Symbol: NF-YC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123691.2 | NP_199139.1 | 1 | 481 | 69 | |
GenBank mRNA | AB493774.1 | - | 1 | 341 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
