Assay Details
| Gene Symbol: GDPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344478.1 | NP_001330568.1 | 2 - 3 | 453 | 78 | |
| NM_123696.5 | NP_199144.3 | 2 - 3 | 457 | 78 | ||
| GenBank mRNA | AK175961.1 | - | 2 - 3 | 381 | 78 | |
More Information
Set Membership: |
|

GDPD3
Genomic Map