Assay Details
| Gene Symbol: ETFBETA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085230.2 | NP_001078699.1 | 4 - 5 | 574 | 77 | |
| NM_001344482.1 | NP_001332757.1 | 4 - 5 | 665 | 77 | ||
| NM_123709.4 | NP_199156.1 | 4 - 5 | 601 | 77 | ||
| GenBank mRNA | AF446867.1 | - | 4 - 5 | 526 | 77 | |
| AK317388.1 | - | 4 - 5 | 563 | 77 | ||
| AY045657.1 | - | 4 - 5 | 575 | 77 | ||
More Information
Set Membership: |
|

ETFBETA
Genomic Map