Assay Details
Gene Symbol: ATAUX2-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123736.3 | NP_199183.1 | 2 - 3 | 761 | 67 | |
GenBank mRNA | AK227549.1 | - | 2 - 3 | 534 | 67 | |
BT005853.1 | - | 2 - 3 | 475 | 67 | ||
L15450.1 | - | 2 - 3 | 475 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
