Assay Details
| Gene Symbol: AGO10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203535.1 | NP_001190464.1 | 4 - 5 | 1257 | 125 | |
| NM_001344505.1 | NP_001331600.1 | 3 - 4 | 1180 | 125 | ||
| NM_001344506.1 | NP_001331599.1 | 3 - 4 | 997 | 125 | ||
| NM_123748.3 | NP_199194.1 | 3 - 4 | 1052 | 125 | ||
More Information
Set Membership: |
|

AGO10
Genomic Map