Assay Details
| Gene Symbol: CBP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085236.2 | NP_001078705.1 | 4 - 5 | 614 | 154 | |
| NM_123787.4 | NP_199233.1 | 3 - 4 | 612 | 154 | ||
| GenBank mRNA | AF140219.1 | - | 3 - 4 | 433 | 154 | |
| AK227850.1 | - | 3 - 4 | 568 | 154 | ||
| BT004813.1 | - | 3 - 4 | 381 | 154 | ||
More Information
Set Membership: |
|

CBP20
Genomic Map