Assay Details
| Gene Symbol: CBP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085236.2 | NP_001078705.1 | 8 - 9 | 926 | 96 | |
| NM_123787.4 | NP_199233.1 | 7 - 8 | 924 | 96 | ||
| GenBank mRNA | AF140219.1 | - | 7 - 8 | 745 | 96 | |
| AK227850.1 | - | 7 - 8 | 880 | 96 | ||
| BT004813.1 | - | 7 - 8 | 693 | 96 | ||
More Information
Set Membership: |
|

CBP20
Genomic Map