Assay Details
Gene Symbol: AT5G44330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123800.1 | NP_199246.1 | 2 - 3 | 405 | 138 | |
GenBank mRNA | DQ056706.1 | - | 2 - 3 | 405 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
