Assay Details
Gene Symbol: VPS2.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125899.1 | NP_001119371.1 | 7 - 8 | 570 | 79 | |
NM_001344559.1 | NP_001332587.1 | 7 - 8 | 551 | 79 | ||
NM_123823.6 | NP_199269.1 | 7 - 8 | 576 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
