Assay Details
Gene Symbol: VPS2.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125899.1 | NP_001119371.1 | 9 - 10 | 758 | 63 | |
NM_123823.6 | NP_199269.1 | 9 - 10 | 764 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
