Assay Details
Gene Symbol: RAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123847.3 | NP_199292.1 | 5 - 6 | 2079 | 75 | |
GenBank mRNA | AF082565.1 | - | 5 - 6 | 1890 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
