Assay Details
Gene Symbol: RAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123847.3 | NP_199292.1 | 8 - 9 | 2872 | 76 | |
GenBank mRNA | AF082565.1 | - | 8 - 9 | 2683 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
